Order Entry
Germany
Orders LinkContactUsLinkComponent
Mobile container spill kit

Mobile container spill kit

Mobile container chemical kit, 128 L, 500×540×900 mm
Catalog # BRDY813883
Supplier:   Brady
Mobile container spill kit
Catalog # BRDY813883
Supplier:  Brady
Mobile container spill kit

Mobile container chemical kit, 128 L, 500×540×900 mm

Price
500,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

These wheeled spill kits are comes in a polyethylene container with hinged lid. Designed for medium sized spills of industrial liquids including oils, water, solvents, coolants and chemical hazardous liquids.

  • Yellow in colour

Container is weather-resistant and protects the sorbents against dirt, moisture and damage. Hinged lid provides easy access to the products. Wheels allow excellent mobility indoors and outdoors.

Specifications
  • Description:
    Mobile container chemical kit
  • Seq:
    02
  • W×D×H:
    500×540×900 mm
  • Absorbency:
    128 L