Trypsin
Trypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol red, 100 ml
Supplier: Biowest
Jump to:
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifications
Specifications
- Pk:100 ml
- Description:Trypsin-EDTA (recombinant) 1X in PBS, without calcium, without magnesium, without phenol red