Orders LinkContactUsLinkComponent
Pipette tips, epT.I.P.S.® Motion
Pipette tips, epT.I.P.S.® Motion
Standard, 40 - 1000 µl, -, 10 racks, 96 each
Catalog # EPPE0030014.480
Supplier:   EPPENDORF
Pipette tips, epT.I.P.S.® Motion
Catalog # EPPE0030014.480
Supplier:  EPPENDORF
Pipette tips, epT.I.P.S.® Motion
Pipette tips, epT.I.P.S.® Motion
Standard, 40 - 1000 µl, -, 10 racks, 96 each
Price
118,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
These pipette tips are specially designed for use on the epMotion 5070 and 5075 Liquid Handling Workstation. They were developed in torsion-resistant, contamination-free racks.
  • High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
  • Tips are optimised for simple automatic ejection in the Liquid Handling Workstation collection vessel
  • Tips can also be integrated into the workstation as a refilling system
  • Refilling tips are delivered in a PET thermoformed tray with sealed lid
  • Replacing the disposable box with an autoclavable aluminum adapter can mean a waste savings of up to 40 percent
The epT.I.P.S. Motion Rack is made up of two components for flexible use in the automated epMotion Liquid Handling Workstation: a colour-coded tray with the epT.I.P.S. filter tips and a disposable box for storing the tray.
Packaging: Packaged in contamination-free racks or in reload trays.
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    40 - 1000 µl
  • Seq:
    66
  • Tip style:
    Standard