Orders LinkContactUsLinkComponent
Feeder cabinets
Feeder cabinets
Feeder cabinets, front load and front dispense with 8 compartments, 1000×400×2000 mm
Catalog # PALBFFCS-1040-SST
Supplier:   Palbam
Feeder cabinets
Catalog # PALBFFCS-1040-SST
Supplier:  Palbam
Feeder cabinets
Feeder cabinets
Feeder cabinets, front load and front dispense with 8 compartments, 1000×400×2000 mm
Price
16 450,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The feeder cabinets support the gowning operation. Made of stainless steel 304 with electropolished finish.
  • Front access doors are used for reloading and for storage purposes
  • The bottom cabinet can be used for storage or disposal
  • Perspex dispenser slots ensure that garments/disposables can be extracted one at a time and do not spill out
Delivery information: Typically available from stock for direct shipment from the supplier. Delivered as shipped assembled or flat packed.
Specifications
  • Description:
    Feeder cabinets, front load and front dispense with 8 compartments
  • Seq:
    02
  • W×D×H:
    1000×400×2000 mm