Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stirrer Heating Mantles WHM, f. round bottom flasks 5000 ml,with integrated power controller,temperature range 450°C, Magnetic stirrer: 50 - 750rpm, 800W, power supply 230V 1 * 1 items
Catalog # WITGDH.WHM12037
Supplier:
witeg Labortechnik
Expand image
Stirrer Heating Mantles WHM, f. round bottom flasks 5000 ml,with integrated power controller,temperature range 450°C, Magnetic stirrer: 50 - 750rpm, 800W, power supply 230V 1 * 1 items
Catalog # WITGDH.WHM12037
Supplier:
witeg Labortechnik