Order Entry
Germany
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

7,00, 20 °C, 10 L
Catalog # APPC272170.1315
Supplier:   PanReac AppliChem
pH buffer solutions
Catalog # APPC272170.1315
Supplier:  PanReac AppliChem
CAS Number:  
pH buffer solutions
pH buffer solutions

7,00, 20 °C, 10 L

CAS Number:  
Price
174,00 €
Log in for availability
1 * 10 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • pH value:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.