Order Entry
Germany
Orders LinkContactUsLinkComponent
 

pH buffer solutions

7,00, 20 °C, 1 L
  APPC272170.1211
pH buffer solutions
  APPC272170.1211
 :  
pH buffer solutions
pH buffer solutions

7,00, 20 °C, 1 L

 :  
 
30,00 €
   
1 * 1 L
 

 

  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • pH value:
    7,00

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.