Ammonium di-Hydrogen Phosphate (Reag. Ph. Eur.) for analysis, ACS 1 * 1.000 g
Catalog # APPC131126.1211
Supplier: PanReac AppliChem
Ammonium di-Hydrogen Phosphate (Reag. Ph. Eur.) for analysis, ACS 1 * 1.000 g
Catalog # APPC131126.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Potassium acetate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Gold Nanourchins (90Nm Diameter, 1 Od, 0 1 * 100 mL
Supplier: Strem Chemicals, Inc.
Ammonium thiocyanate 98% ACS
Supplier: Thermo Fisher Scientific
Sodium acetate, anhydrous 99%
Supplier: Thermo Fisher Scientific
1-Heptane Sulfonic Acid Sodium Salt 1-hydrate for HPLC 1 * 25 g
Supplier: PanReac AppliChem
Tin(Ii) Chloride, Anhydrous, 98% 1 * 500 g
Supplier: Strem Chemicals, Inc.
Starch, soluble (from potato) Reag. USP, Ph. Eur. analytical reagent
Supplier: PanReac AppliChem
di-Potassium Hydrogen Phosphate anhydrous (BP, Ph. Eur.) pure, pharma grade 1 * 1.000 g
Supplier: PanReac AppliChem
Copper(II) oxide ≥97.5%, powder, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Diammonium hydrogen orthophosphate ≥98%
Supplier: Thermo Fisher Scientific
Benzyl alcohol ACS
Supplier: Thermo Fisher Scientific