Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Catalog # APPC131007.1212
Supplier: PanReac AppliChem
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Catalog # APPC131007.1212
Supplier: PanReac AppliChem
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 5 L
Supplier: PanReac AppliChem
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetonitrile LC-MS 2.5L 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals