Mesenchymal stem cell differentiation medium
Mesenchymal Stem Cell Chondrogenic Differentiation Medium, without inducers (ready-to-use), 100 ml, 100 mL
Catalog # C-28014
Marketsource
Supplier: PROMOCELL
Mesenchymal stem cell differentiation medium
Catalog # C-28014
Supplier: PROMOCELL
Marketsource
CAS Number:
Jump to:
Product Details & Documents
PromoCell Mesenchymal Stem Cell Differentiation Media efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively.
Specifications
Specifications
- Pk:100 mL
- Description:Mesenchymal Stem Cell Chondrogenic Differentiation Medium, without inducers (ready-to-use), 100 ml



