Inserts
E 416 insert 1/2, for 6 one-or-two part specula, 7 holders, for use in upper and lower baskets, G 7882, G 7892, G 7882 CD, G 7835/36 CD, 178×279×157 mm
Catalog # MIEL03888650
Supplier: MIELE
Jump to:
Product Details & Documents
Specifications
- Description:E 416 insert 1/2, for 6 one-or-two part specula, 7 holders, for use in upper and lower baskets
- Seq:53
- W×D×H:178×279×157 mm
- For:G 7882, G 7892, G 7882 CD, G 7835/36 CD