G‐Biosciences Colloidal Blue Stain is a mass spectrometry compatible stain that offers nanogram sensitive detection of proteins. The stain offers ten times more sensitivity compared to classical Coomassie R‐ or G‐250 detection. The Colloidal Blue St 1 * 1 L
Catalog # GENO786-500
Lieferant: G-Biosciences
Frequently Bought Together