Vancomycin hydrochloride
Bottle
Catalog # GENORC-205
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Vancomycin is a glycopeptide antibiotic that exerts its action by preventing the transfer and addition of the muramylpentapeptides that comprise the peptidoglycan. Vancomycin is often used synergistically with cefotaxime or carbenicillin against Agrobacterium.
- Molecular Biology Grade
- Tested against both sensitive and resistant cells
- Potency of greater than 1000 mcg/mg
Synonyms: (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
- CAS Number:1404-93-9
- Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
- Molecular Weight:1485,71 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



