Order Entry
Germany
Orders LinkContactUsLinkComponent
Turbidity standards, Stablcal®

Turbidity standards, Stablcal®

Stablcal® stabilised formazine standard, 0,06 NTU, 100 ml
Catalog # 708-0085
Supplier:   Hach
Turbidity standards, Stablcal®
Catalog # 708-0085
Supplier:  Hach
CAS Number:  
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®

Stablcal® stabilised formazine standard, 0,06 NTU, 100 ml

CAS Number:  
Price
93,60 €
Log in for availability
1 * 100 mL
Product Details & Documents

Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.

  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results

Documents

Specifications

Specifications

  • Pk:
    100 ml
  • Turbidity:
    0,06 NTU
  • Description:
    Stablcal® stabilised formazine standard

Specifications