Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Copper standard solution

Copper standard solution (NIST), 1000 mg/l as Cu, 100 mL
Catalog # HACH259342
Supplier:   Hach
Copper standard solution
Catalog # HACH259342
Supplier:  Hach
CAS Number:  
Copper standard solution

Copper standard solution (NIST), 1000 mg/l as Cu, 100 mL

CAS Number:  
Price
53,70 €
Log in for availability
1 * 100 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Standard solution for accuracy checks in copper determinations.

Specifications

Specifications

  • Concentration:
    1000 mg/l as Cu
  • Pk:
    100 mL
  • Description:
    Copper standard solution (NIST)

Specifications