Order Entry
Germany
Orders LinkContactUsLinkComponent
pH Buffer solutions

pH Buffer solutions

Buffer solution, 7,00, Colourless, 25 mL
Catalog # HACH1222220
Supplier:   Hach
pH Buffer solutions
Catalog # HACH1222220
Supplier:  Hach
CAS Number:  
pH Buffer solutions
pH Buffer solutions

Buffer solution, 7,00, Colourless, 25 mL

CAS Number:  
Price
21,60 €
Log in for availability
1 * 25 mL
Product Details & Documents

For calibration of pH meters.

Accuracy: pH is ±0,02 at 25 °C

Certifications: Traceable to NIST.

Caution: Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

Specifications

Specifications

  • Pk:
    25 mL
  • Colour:
    Colourless
  • Description:
    Buffer solution
  • pH value:
    7,00

Specifications