Order Entry
Germany
Orders LinkContactUsLinkComponent
Phenol red solution pH-indicator (pH 6,5 - 8,5)

Phenol red solution pH-indicator (pH 6,5 - 8,5)

Dropping bottle, self-contained, 50 ml
Catalog # HACH21126
Supplier:   Hach
Phenol red solution pH-indicator (pH 6,5 - 8,5)
Catalog # HACH21126
Supplier:  Hach
CAS Number:  143-74-8
Phenol red solution pH-indicator (pH 6,5 - 8,5)
Phenol red solution pH-indicator (pH 6,5 - 8,5)

Dropping bottle, self-contained, 50 ml

CAS Number:  143-74-8
Price
20,70 €
Log in for availability
1 * 50 mL
Product Details & Documents

Phenol Red Indicator Solution provides an easy-to-use colorimetric method for determining pH levels for neutral and slightly alkaline waters in the range of 6.5 to 8.5 units. This method uses a color wheel and is especially useful for testing swimming pools and spas.

In addition to these applications, Phenol Red is also used for monitoring ground and river water quality, making it a versatile tool for various environmental water quality testing needs.

Color range: pH 6.5 - 8.5

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
Specifications

Specifications

  • Pack type:
    Dropping bottle, self-contained
  • Pk:
    50 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.