Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hanus Reagent 0.1 mol/l (0.2N) for volumetric analysis 1 * 1.000 mL
Catalog # APPC281572.1611
Supplier:
PanReac AppliChem
Expand image
Hanus Reagent 0.1 mol/l (0.2N) for volumetric analysis 1 * 1.000 mL
Catalog # APPC281572.1611
Supplier:
PanReac AppliChem