Sodium hypochlorite solution 5% (w/w), Technical Grade
Plastic container, 5 L
Supplier: PanReac AppliChem
Sodium hypochlorite solution 5% (w/w), Technical Grade
Catalog # APPC212297.1214
Supplier: PanReac AppliChem
CAS Number: 7681-52-9
Jump to:
Product Details & Documents
- CAS Number:7681-52-9
- Formula:ClNaO
- MDL Number:MFCD00011120
- Molecular Weight:74.44 g/mol
- Storage Temperature:Ambient
- UN:1791
- ADR:8,III
Documents
Specifications
Specification Test Results
- Assay (iodometr.):5% ± 0.5% (At the moment of the batch analysis)
Specifications
- Pack type:Plastic container
- Pk:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



