Nitric Acid 69% for trace metal analysis (ppb) 1 * 500 mL
Catalog # APPC721037.0010
Supplier: PanReac AppliChem
Nitric Acid 69% for trace metal analysis (ppb) 1 * 500 mL
Catalog # APPC721037.0010
Supplier: PanReac AppliChem
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Latex Gloves
Supplier: VWR Collection