Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Sodium carbonate / hydrogen carbonate eluent for IC 3.2 mmol/l Na2CO3 + 1.0 mmol/l NaHCO3 in water 1 * 2.500 mL
KRAF22841.4000
:
Bernd Kraft
Sodium carbonate / hydrogen carbonate eluent for IC 3.2 mmol/l Na2CO3 + 1.0 mmol/l NaHCO3 in water 1 * 2.500 mL
KRAF22841.4000
:
Bernd Kraft