ContactUsLinkComponent
FLUORESCENT DYE 633-I STREPTAVIDIN FLUORESCENCE DYE 633-I WITH STREPTAVIDIN CONJUGATE. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 63 3-I IS WELL SEPARATED FROM THAT OF OTHER COMMONLY USED RED FLUOROPHORES, SUCH AS TAMRA, TEXAS RED, ALEXA FLUOR 594, F 1 * 200 µG
Catalog # ABNOU0295
Supplier:  Abnova
FLUORESCENT DYE 633-I STREPTAVIDIN FLUORESCENCE DYE 633-I WITH STREPTAVIDIN CONJUGATE. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 63 3-I IS WELL SEPARATED FROM THAT OF OTHER COMMONLY USED RED FLUOROPHORES, SUCH AS TAMRA, TEXAS RED, ALEXA FLUOR 594, F 1 * 200 µG
Catalog # ABNOU0295
Supplier:  Abnova