ContactUsLinkComponent
Coulometer for easy and fast low level water determinations in liquid, solid and gaseous samples. With four MSB connections for 800 Dosino for the addition of samples, standard and auxiliary solutions. Including titration cell with diaphragm and 801 1 * 1 items
Catalog # METR2.851.0020
Supplier:  METROHM
Coulometer for easy and fast low level water determinations in liquid, solid and gaseous samples. With four MSB connections for 800 Dosino for the addition of samples, standard and auxiliary solutions. Including titration cell with diaphragm and 801 1 * 1 items
Catalog # METR2.851.0020
Supplier:  METROHM
Frequently Bought Together