ContactUsLinkComponent
Precellys Evolution Super Homogenizer incl. 24 x 0.5/2.0 ml tubes option High Performance Beat Beater for fast, effective and reproducible homogenisation of samples in 0.5/2.0 ml, 7 ml*, or 15 ml* tubes without any cross-contaminations. Best grinding 1 * 1 items
Catalog # 432-0275
Supplier:  Bertin Instruments
undefined
Precellys Evolution Super Homogenizer incl. 24 x 0.5/2.0 ml tubes option High Performance Beat Beater for fast, effective and reproducible homogenisation of samples in 0.5/2.0 ml, 7 ml*, or 15 ml* tubes without any cross-contaminations. Best grinding 1 * 1 items
Catalog # 432-0275
Supplier:  Bertin Instruments
Frequently Bought Together