Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Human PD-L1/B7-H1 Protein (ECD, His Tag), HPLC-verified 1 * 100 µG
Catalog # SINO10084-H08H-100
Supplier:
Sino Biological
Expand image
Recombinant Human PD-L1/B7-H1 Protein (ECD, His Tag), HPLC-verified 1 * 100 µG
Catalog # SINO10084-H08H-100
Supplier:
Sino Biological