ContactUsLinkComponent
cabinet for chemicals, tall cabinet with 3 wing doors right hinged, model line c-line, model cs.195.054.mh.3wdr, version horizontal dividing wall dimensions: exterior: wxdxh (mm) 545 x 520 x 1950 interior: wxdxh (mm) 486 x 495 x 595 body: ral 7016, y 1 * 1 items
Catalog # 135-1941
Supplier:  asecos
cabinet for chemicals, tall cabinet with 3 wing doors right hinged, model line c-line, model cs.195.054.mh.3wdr, version horizontal dividing wall dimensions: exterior: wxdxh (mm) 545 x 520 x 1950 interior: wxdxh (mm) 486 x 495 x 595 body: ral 7016, y 1 * 1 items
Catalog # 135-1941
Supplier:  asecos
Frequently Bought Together