Septa caps
Open top septum cap, bonded 1,14 mm, PTFE-lined silicon septa, 53-400
Catalog # EPSCS297-5300
Supplier: EP SCIENTIFIC
Jump to:
Product Details & Documents
PP, white.
Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.
Specifications
- Thread:53-400
- Description:Open top septum cap, bonded 1,14 mm, PTFE-lined silicon septa
- Seq:07
- Cat. No.:EPSCS297-5300
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #TypeForØAvailabilityPrice
Product Information
- Catalog #TypeForØAvailabilityPrice
Product Information
- Catalog #TypeForØAvailabilityPrice
- EPSC288-0022BPSilicone, PTFE-lined24-214 VOA vials24 mmLog in for availability1 * 2.000 ST - 2 430,00 €
Product Information
- Catalog #TypeForØAvailabilityPrice
- EPSCS24-400-S2Silicone, PTFE-lined24-214 VOA vials24 mmLog in for availability1 * 144 ST - 287,00 €