Orders LinkContactUsLinkComponent
Cleanroom boot covers, Kimtech™ A5
Cleanroom boot covers, Kimtech™ A5
Wrap-around vinyl foot, White, S/M, +
Catalog # 113-8197
   Waste Reducing
Supplier:   Ansell
Cleanroom boot covers, Kimtech™ A5
Catalog # 113-8197
Supplier:  Ansell
   Waste Reducing
Cleanroom boot covers, Kimtech™ A5
Cleanroom boot covers, Kimtech™ A5
Wrap-around vinyl foot, White, S/M, +
Price
832,00 €
Log in for availability
1 * 100 PAAR
Product Details & Documents
Sterile Kimtech™ A5 cleanroom boot covers are made of SMS (Spunbond Melt-blown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
  • Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards
  • Tunnelled elastic and bound seams
  • Two extra-long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
Certifications: PPE Cat I (EU) 2016/425
Packaging: Vacuum packed in pairs for added sterility assurance.
Specifications
  • Shoe size:
    S/M
  • Sterile:
    +
  • Colour:
    White
  • Description:
    Wrap-around vinyl foot
  • Seq:
    05
  • Cat. No.:
    113-8197
  • Cleanroom class:
    100/ISO 5
  • Garment application:
    Cleanroom
  • Environmentally Preferable:
    Waste Reducing
Complementary Products
Product Information
  • Catalog #
    Description
    Sterile
    Clothing Size
    Colour
    Length
    Availability
    Price
  • 113-1199
    Without hood
    +
    M
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-1210
    Without hood
    +
    L
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-1198
    Without hood
    +
    S
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-1211
    Without hood
    +
    XL
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-1212
    Without hood
    +
    XXL
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-1213
    Without hood
    +
    3XL
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-1219
    Without hood
    +
    4XL
    White
    Log in for availability
    1 * 25 ST - 581,00 €
  • 113-7732
    With integrated mask
    +
    01
    Log in for availability
    1 * 75 ST - 754,00 €
  • 114-9642
    Standard
    +
    03
    Log in for availability
    1 * 100 ST - 471,00 €
  • 113-7733
    457 mm
    +
    White
    Log in for availability
    1 * 100 PAAR - 520,00 €