Disposable filters, Optimizer® DEV and DI
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
- Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
- O-ringless connections avoid potential sources of contamination
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, core and sleeve: HDPE; Disposable shell: PP
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,8 bar at 25 °C
Maximum reverse differential pressure: 2,8 bar at 25 °C
Articles may not be available in all countries. Please contact your local Avantor office.
- Filtration area:3600 cm²
- Pore size:0,05 µm
- Type:DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm
- Seq:32



