ContactUsLinkComponent
C18G x 3µm particle size x 300Å pore size, 10mm length x 150µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C184-03G15K
C18G x 3µm particle size x 300Å pore size, 10mm length x 150µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C184-03G15K
Frequently Bought Together