ContactUsLinkComponent
C18Q x 3µm particle size x 100Å pore size, 10mm length x 4.0mm ID, Stainless steel ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C183-03G04S
C18Q x 3µm particle size x 100Å pore size, 10mm length x 4.0mm ID, Stainless steel ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C183-03G04S
Frequently Bought Together