ContactUsLinkComponent
C18H x 3µm particle size x 120Å pore size, 10mm length x 300µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C182-03G30K
C18H x 3µm particle size x 120Å pore size, 10mm length x 300µm ID, PEEKsil ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C182-03G30K
Frequently Bought Together