ContactUsLinkComponent
C18H x 3µm particle size x 120Å pore size, 10mm length x 2.1mm ID, Stainless steel ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C182-03G02S
C18H x 3µm particle size x 120Å pore size, 10mm length x 2.1mm ID, Stainless steel ProteCol HPLC Guard Column PK3 1 * 1 items
Catalog # SGEA2C182-03G02S
Frequently Bought Together