ContactUsLinkComponent
INPP5E (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETE CT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 SET 1 * 1 SET
Catalog # ABNOH00056623-PW1
Supplier:  Abnova
INPP5E (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETE CT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 SET 1 * 1 SET
Catalog # ABNOH00056623-PW1
Supplier:  Abnova