ContactUsLinkComponent
TNFRSF17 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DE TECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 SET 1 * 1 SET
Catalog # ABNOH00000608-PW2
Supplier:  Abnova
TNFRSF17 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DE TECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 SET 1 * 1 SET
Catalog # ABNOH00000608-PW2
Supplier:  Abnova