Nitric acid 68%, TECHNICAL
Plastic bottle, 2,5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Nitric acid solution
- CAS Number:7697-37-2
- Formula:HNO₃
- MDL Number:MFCD00011349
- Molecular Weight:63.01 g/mol
- Merck Index:13,06608
- Boiling Pt:120.5 °C (1013 hPa)
- Melting Pt:–42 °C
- Density:1.4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-714-2
- ADR:8 (5.1),II
- :Density: 1.51 g/cm³ (20 °C)
- UN:2031
Specifications
Specification Test Results
- Assay:68 - 70 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L



