Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE DRAWER ET-M 12 LITRES 1 * 4 items
Catalog # MULCLIGHT_DRAWER_M
Supplier:
MULTIROIR
Expand image
STORAGE DRAWER ET-M 12 LITRES 1 * 4 items
Catalog # MULCLIGHT_DRAWER_M
Supplier:
MULTIROIR