Orders LinkContactUsLinkComponent
Sample spoons, mini, SP Bel-Art
Sample spoons, mini, SP Bel-Art
1,0 ml, PS
Catalog # BELAF367210100
Sample spoons, mini, SP Bel-Art
Catalog # BELAF367210100
Sample spoons, mini, SP Bel-Art
Sample spoons, mini, SP Bel-Art
1,0 ml, PS
Price
60,90 €
Log in for availability
1 * 25 ST
Product Details & Documents
PS, white.
  • Disposable, made from white high-impact PS
  • Ideal for collecting specific small volumes of powders and other dry materials
  • 0,10 / 0,50 / 1,0 ml models have a rounded bowl for easier emptying of the spoon and a thinner neck that minimises extra material collection
Documents
Specifications
  • Capacity:
    1,0 ml
  • Material:
    PS
  • Seq:
    05