F-35-6-30 Fixed Angle Rotor with Lid Max. Capacity: 6 x 15/50 ml Falcon tubes Max. Speed: 7,830 rpm/7,197 g 1 * 1 items
Catalog # EPPE5427739.009
F-35-6-30 Fixed Angle Rotor with Lid Max. Capacity: 6 x 15/50 ml Falcon tubes Max. Speed: 7,830 rpm/7,197 g 1 * 1 items
Catalog # EPPE5427739.009