Orders LinkContactUsLinkComponent
Objectives for PSM-1000 microscopes
Objectives for PSM-1000 microscopes
ELWD plan apo objective (WD = 13 mm)
Catalog # MOTI1101001700062
Supplier:   MOTIC
Objectives for PSM-1000 microscopes
Catalog # MOTI1101001700062
Supplier:  MOTIC
Objectives for PSM-1000 microscopes
ELWD plan apo objective (WD = 13 mm)
Price
1 820,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    50×/0,55
  • Description:
    ELWD plan apo objective (WD = 13 mm)
  • Seq:
    109