Orders LinkContactUsLinkComponent
Eyepieces
Eyepieces
N-WF ESD eyepiece
Catalog # MOTI1101001402731
Supplier:   MOTIC
Eyepieces
Catalog # MOTI1101001402731
Supplier:  MOTIC
Eyepieces
Eyepieces
N-WF ESD eyepiece
Price
118,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    15× (16 mm)
  • Description:
    N-WF ESD eyepiece
  • Seq:
    102