ContactUsLinkComponent
qPCR detection of Mycoplasma species haemofelis and haemocanis. Contains: Primer and probe mix, Reverse Transcription target specifi c primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-M.HFC
Supplier:  PRIMERDESIGN
qPCR detection of Mycoplasma species haemofelis and haemocanis. Contains: Primer and probe mix, Reverse Transcription target specifi c primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-M.HFC
Supplier:  PRIMERDESIGN