ContactUsLinkComponent
qPCR detection of Human Polyomavirus 9. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, E ndogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HPYV9
Supplier:  PRIMERDESIGN
qPCR detection of Human Polyomavirus 9. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, E ndogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HPYV9
Supplier:  PRIMERDESIGN