ContactUsLinkComponent
qPCR detection of Klebsiella oxytoca. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viru ses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-K.OXY
Supplier:  PRIMERDESIGN
qPCR detection of Klebsiella oxytoca. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viru ses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-K.OXY
Supplier:  PRIMERDESIGN