Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Multi layer adhesive floor mats

White, 610×914 mm, 0,036 mm
  PURUPM243638W
 :   PURUS
Multi layer adhesive floor mats
  PURUPM243638W
 :  PURUS
Multi layer adhesive floor mats
Multi layer adhesive floor mats

White, 610×914 mm, 0,036 mm

 
448,00 €
   
1 * 8 ST
 

A choice of either 30 or 60 layer adhesive entrance mats are manufactured from low density polyethylene. The mats control contamination by removing debris from the soles of shoes and the wheels of transport containers. When one layer is dirty, simply remove by the numbered corner tab, leaving a fresh layer ready for use.

  • Antimicrobial agent protects against the growth of bacteria and fungi
  • Non-slip, low profile, individually numbered layers
  • Reduced maintenance and cleaning costs

Colour: Clear, white, grey or blue

 
  • Colour:
    White
  • L×W:
    610×914 mm
  • Thickness:
    0,036 mm