Safety Posters
COSHH guidance poster
Catalog # 116-0424
Supplier: SAFETY FIRST AID GROUP
Jump to:
Product Details & Documents
The Chemical Spills poster has clear step by step instructions on how to react in case of a chemical spill and how to provide treatment to someone that has been affected. The poster is designed for both first aiders and non first aiders with the essential first aid and safety information.
- Fully coloured illustrations and clear step by step instructions with easy to follow text
- Compiled by qualified health and safety practitioners
- Encapsulated finish for superior durability
The COSHH poster is designed to help protect employees and avoid costly accidents. User friendly text and clear instructions show employees how to prevent accidents and injury whilst maintaining a healthy working environment.
Specifications
- Description:COSHH guidance poster
- Seq:02