Orders LinkContactUsLinkComponent
Gowning racks, wall-mounted
Gowning racks, wall-mounted
Wall-mounted garment rack + 32 hangers, 1709×468×470 mm
Catalog # 132-0991
Supplier:   Palbam
Gowning racks, wall-mounted
Catalog # 132-0991
Supplier:  Palbam
Gowning racks, wall-mounted
Gowning racks, wall-mounted
Wall-mounted garment rack + 32 hangers, 1709×468×470 mm
Price
2 190,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Wall-mounted gownracks in stainless steel 304 with electropolished finish.
  • Gowning racks include stainless steel hangers
  • The design aligns the hangers, keeping an orderly appearance and ensuring the hangers do not touch each other
Delivery information: Typically available from stock for direct shipment from the supplier. Shipped flat-packed with assembly instructions.
Specifications
  • Description:
    Wall-mounted garment rack + 32 hangers
  • Seq:
    04
  • W×D×H:
    1709×468×470 mm