ContactUsLinkComponent
Library quant Primer(Illumina) ABI Prism KAPA PRIMER & MIX (ILLUMINA - ABI PRISM)KAPA Library Quant Primer Premix and qPCR MasterMix Illumina - ABI Prism (ROX High). Components of the KAPA Library Quant Illumina ABI Prism kit for the quantification f 1 * 1 KIT
Catalog # KAPBKK4933
Supplier:  KAPA BIOSYSTEMS
Library quant Primer(Illumina) ABI Prism KAPA PRIMER & MIX (ILLUMINA - ABI PRISM)KAPA Library Quant Primer Premix and qPCR MasterMix Illumina - ABI Prism (ROX High). Components of the KAPA Library Quant Illumina ABI Prism kit for the quantification f 1 * 1 KIT
Catalog # KAPBKK4933
Supplier:  KAPA BIOSYSTEMS