Order Entry
Germany
Orders LinkContactUsLinkComponent
Spray guns

Spray guns

Spray gun, natural, with 6,35 mm FNPT connection
Catalog # ENTE421-32-11
Supplier:   ENTEGRIS
Spray guns
Catalog # ENTE421-32-11
Supplier:  ENTEGRIS
Spray guns

Spray gun, natural, with 6,35 mm FNPT connection

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Made of high purity PFA and PTFE (all wetted parts).

  • With 4,8 mm orifice and 6,35 and 12,7 mm FNPT inlet connections
  • Max. pressure 5,52 bar
  • Coloured caps enable visual chemical differentiation at a glance
Specifications
  • Description:
    Spray gun, natural, with 6,35 mm FNPT connection
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.