Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Tobramycin

1 g
Catalog # J66040.03
Tobramycin
Catalog # J66040.03
CAS Number:  32986-56-4
Tobramycin
Tobramycin

1 g

CAS Number:  32986-56-4
Price
90,50 €
Log in for availability
1 * 1 g
Product Details & Documents

Tobramycin primarily targets gram negative bacteria.

  • White to Off-White powder

Soluble in water

Synonyms:  (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
  • CAS Number:32986-56-4
  • Formula:C₁₈H₃₇N₅O₉
  • MDL Number:MFCD00077885
  • Molecular Weight:467.51 g/mol
  • Merck Index:14,9490
  • Storage Temperature:Refrigerator
  • EINECS:251-322-5
Specifications

Specifications

  • Pk:
    1 g

Specifications